ENA429358772 Enamine

(4S)-2-(3-hydroxyphenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD18447511
  • Cas Number : 1217529-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186480
Availability details for
Quantity
  • Availability
  • Add to Cart