ENA429358773 Enamine

4-[4-(methylsulfanyl)phenyl]pyrimidin-2-amine

  • MDL number MFCD07381853
  • Cas Number : 1206108-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186481

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186481
Availability details for
Quantity
  • Availability
  • Add to Cart