ENA429358775 Enamine

4-(2,4-dichlorophenyl)pyrimidin-2-amine

  • MDL number MFCD04050240
  • Cas Number : 828273-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186483
Availability details for
Quantity
  • Availability
  • Add to Cart