ENA429358776 Enamine

4-(4-chloro-2-fluorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD09996851
  • Cas Number : 1010902-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186484
Availability details for
Quantity
  • Availability
  • Add to Cart