ENA429358778 Enamine

5-(2-methoxyphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00981010
  • Cas Number : 28004-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186486

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186486
Availability details for
Quantity
  • Availability
  • Add to Cart