ENA429358785 Enamine

3',4'-dihydro-1'H-spiro[cyclohexane-1,2'-quinazoline]-4'-one

  • MDL number MFCD02029491
  • Cas Number : 950-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186493

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186493
Availability details for
Quantity
  • Availability
  • Add to Cart