ENA429358790 Enamine

2-[2-(1H-pyrrol-1-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD02665538
  • Cas Number : 383142-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186498

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186498
Availability details for
Quantity
  • Availability
  • Add to Cart