ENA429358795 Enamine

1'-methyl-3,5,6,7-tetrahydrospiro[imidazo[4,5-c]pyridine-4,4'-piperidine]

  • MDL number MFCD09749716
  • Cas Number : 65092-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186503
Availability details for
Quantity
  • Availability
  • Add to Cart