ENA429358800 Enamine

2-[(3S)-2,5-dioxo-2,3,4,5-tetrahydro-1H-1,4-benzodiazepin-3-yl]acetic acid

  • MDL number MFCD10007533
  • Cas Number : 1217715-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186508
Availability details for
Quantity
  • Availability
  • Add to Cart