ENA429358802 Enamine

1'-(propan-2-yl)-3,5,6,7-tetrahydrospiro[imidazo[4,5-c]pyridine-4,4'-piperidine]

  • MDL number MFCD09749784
  • Cas Number : 929809-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186510

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186510
Availability details for
Quantity
  • Availability
  • Add to Cart