ENA429358807 Enamine

4-{3H,4H,5H,6H,7H-imidazo[4,5-c]pyridin-4-yl}pyridine

  • MDL number MFCD07384682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186515
Availability details for
Quantity
  • Availability
  • Add to Cart