ENA429358811 Enamine

2-(1H-pyrrol-1-yl)-1,3-benzothiazole-6-carboxylic acid

  • MDL number MFCD01764668
  • Cas Number : 251457-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186519
Availability details for
Quantity
  • Availability
  • Add to Cart