ENA429358817 Enamine

6-(1H-1,2,3,4-tetrazol-1-yl)hexanoic acid

  • MDL number MFCD11868191
  • Cas Number : 117027-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186525
Availability details for
Quantity
  • Availability
  • Add to Cart