ENA429358820 Enamine

4-chloro-2-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD09049884
  • Cas Number : 926189-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186528

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186528
Availability details for
Quantity
  • Availability
  • Add to Cart