ENA429358821 Enamine

3-(2,4-dioxo-1,2,3,4-tetrahydroquinazolin-3-yl)propanoic acid

  • MDL number MFCD05027536
  • Cas Number : 82603-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186529
Availability details for
Quantity
  • Availability
  • Add to Cart