ENA429358823 Enamine

(3R)-3-(2-methylpropyl)-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-2,5-dione

  • MDL number MFCD15730797
  • Cas Number : 1243086-77-2
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-186531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186531
Availability details for
Quantity
  • Availability
  • Add to Cart