ENA429358832 Enamine

3-(5-bromo-1H-indol-1-yl)propanoic acid

  • MDL number MFCD09855471
  • Cas Number : 18108-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186540
Availability details for
Quantity
  • Availability
  • Add to Cart