ENA429358835 Enamine

4-{5,7-dioxo-5H,6H,7H-pyrrolo[3,4-b]pyridin-6-yl}butanoic acid

  • MDL number MFCD11108868
  • Cas Number : 57111-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186543
Availability details for
Quantity
  • Availability
  • Add to Cart