ENA429358836 Enamine

2-{1,3-dioxo-1H,2H,3H-pyrrolo[3,4-c]pyridin-2-yl}acetic acid

  • MDL number MFCD03452532
  • Cas Number : 36239-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186544
Availability details for
Quantity
  • Availability
  • Add to Cart