ENA429358839 Enamine

2-methanesulfonamido-1,3-benzothiazole-6-carboxylic acid

  • MDL number MFCD09849602
  • Cas Number : 929970-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186547
Availability details for
Quantity
  • Availability
  • Add to Cart