ENA429358843 Enamine

6-hydroxy-4-methyl-1-(2-methylpropyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD03307455
  • Cas Number : 88009-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186551
Availability details for
Quantity
  • Availability
  • Add to Cart