ENA429358846 Enamine

3-[(4-methoxyphenyl)methyl]-2-sulfanylidene-1,3-thiazolidin-4-one

  • MDL number MFCD07344274
  • Cas Number : 34396-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186554
Availability details for
Quantity
  • Availability
  • Add to Cart