ENA429358847 Enamine

N-(2,3,4,9-tetrahydro-1H-carbazol-1-ylidene)hydroxylamine

  • MDL number MFCD00623744
  • Cas Number : 91391-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186555

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186555
Availability details for
Quantity
  • Availability
  • Add to Cart