ENA429358848 Enamine

8-methyl-2,3,4,9-tetrahydro-1H-carbazol-1-one

  • MDL number MFCD16090101
  • Cas Number : 3449-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186556
Availability details for
Quantity
  • Availability
  • Add to Cart