ENA429358852 Enamine

3-(dimethylsulfamoyl)-4-methylbenzoic acid

  • MDL number MFCD09042889
  • Cas Number : 926248-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186560
Availability details for
Quantity
  • Availability
  • Add to Cart