ENA429358855 Enamine

3-(oxolan-2-ylmethyl)-2-sulfanylidene-1,3-thiazolidin-4-one

  • MDL number MFCD00178440
  • Cas Number : 42487-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186563
Availability details for
Quantity
  • Availability
  • Add to Cart