ENA429358857 Enamine

1-(6-oxo-1,6-dihydropyridazin-3-yl)piperidine-4-carboxylic acid

  • MDL number MFCD20535474
  • Cas Number : 919536-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186565
Availability details for
Quantity
  • Availability
  • Add to Cart