ENA429358862 Enamine

methyl 5-amino-[1,2,4]triazolo[4,3-a]pyrimidine-6-carboxylate

  • MDL number MFCD20535470
  • Cas Number : 1352504-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186570

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186570
Availability details for
Quantity
  • Availability
  • Add to Cart