ENA429358863 Enamine

6-(4-methylpiperidin-1-yl)pyridazin-3-ol

  • MDL number MFCD15194894
  • Cas Number : 756901-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186572

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186572
Availability details for
Quantity
  • Availability
  • Add to Cart