ENA429358869 Enamine

2-(3-fluorophenyl)-1-methyl-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD07379673
  • Cas Number : 1266973-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186578
Availability details for
Quantity
  • Availability
  • Add to Cart