ENA429358870 Enamine

3-(2,3-dihydro-1-benzofuran-5-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD26523053
  • Cas Number : 1491131-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186579
Availability details for
Quantity
  • Availability
  • Add to Cart