ENA429358871 Enamine

5-(2-methylpropyl)-2-(1H-pyrrol-1-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD19691830
  • Cas Number : 1322605-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186580
Availability details for
Quantity
  • Availability
  • Add to Cart