ENA429358875 Enamine

6-amino-3-methyl-3,4-dihydro-1,2,3-benzotriazin-4-one

  • MDL number MFCD26523096
  • Cas Number : 99055-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186585

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186585
Availability details for
Quantity
  • Availability
  • Add to Cart