ENA429358878 Enamine

5-benzyl-2-methyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD24038557
  • Cas Number : 1429901-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186588

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186588
Availability details for
Quantity
  • Availability
  • Add to Cart