ENA429358881 Enamine

1,3-dimethyl-1H,4H,6H,7H-pyrano[4,3-b]pyrrole-4,6-dione

  • MDL number MFCD14715933
  • Cas Number : 86814-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186591
Availability details for
Quantity
  • Availability
  • Add to Cart