ENA429358885 Enamine

2-amino-5-(2-methylpropyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD22375195
  • Cas Number : 1383626-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186595

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186595
Availability details for
Quantity
  • Availability
  • Add to Cart