ENA429358887 Enamine

6-bromo-2H,3H-[1,3]oxazolo[4,5-b]pyridin-2-one

  • MDL number MFCD08689590
  • Cas Number : 21594-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186597
Availability details for
Quantity
  • Availability
  • Add to Cart