ENA429358893 Enamine

methyl 2-amino-5-propyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD23135600
  • Cas Number : 649736-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186603

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186603
Availability details for
Quantity
  • Availability
  • Add to Cart