ENA429358895 Enamine

2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD22375179
  • Cas Number : 1017273-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186605
Availability details for
Quantity
  • Availability
  • Add to Cart