ENA429358899 Enamine

1-(1H-pyrrol-1-yl)cyclohexane-1-carboxylic acid

  • MDL number MFCD14281661
  • Cas Number : 1031596-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186609
Availability details for
Quantity
  • Availability
  • Add to Cart