ENA429358902 Enamine

2-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)acetic acid

  • MDL number MFCD01790620
  • Cas Number : 127264-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186612
Availability details for
Quantity
  • Availability
  • Add to Cart