ENA429358907 Enamine

3-(6-chloro-1H-indol-1-yl)propanoic acid

  • MDL number MFCD11212281
  • Cas Number : 1094641-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186618

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186618
Availability details for
Quantity
  • Availability
  • Add to Cart