ENA429358910 Enamine

2-(4-methylphenyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD01361064
  • Cas Number : 67189-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186621
Availability details for
Quantity
  • Availability
  • Add to Cart