ENA429358913 Enamine

5-amino-1-(2,4-difluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD03407405
  • Cas Number : 918405-21-7
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-186624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186624
Availability details for
Quantity
  • Availability
  • Add to Cart