ENA429358914 Enamine

4-(1H-1,2,3,4-tetrazol-1-yl)butanoic acid

  • MDL number MFCD11868190
  • Cas Number : 92614-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186625

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186625
Availability details for
Quantity
  • Availability
  • Add to Cart