ENA429358926 Enamine

2-[(octahydro-1H-quinolizin-1-ylmethyl)amino]ethan-1-ol

  • MDL number MFCD24672988
  • Cas Number : 1692839-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186637
Availability details for
Quantity
  • Availability
  • Add to Cart