ENA429358927 Enamine

methyl 2-(2-acetamido-1,3-thiazol-4-yl)acetate

  • MDL number MFCD09881014
  • Cas Number : 126781-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186638
Availability details for
Quantity
  • Availability
  • Add to Cart