ENA429358935 Enamine

2-[(2-aminophenyl)formamido]butanoic acid

  • MDL number MFCD14597910
  • Cas Number : 1248246-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186648
Availability details for
Quantity
  • Availability
  • Add to Cart