ENA429358943 Enamine

3-[(2-amino-9H-purin-6-yl)amino]propanoic acid

  • MDL number MFCD26522937
  • Cas Number : 1573548-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186656
Availability details for
Quantity
  • Availability
  • Add to Cart