ENA429358945 Enamine

2-[1-(6-chloropyridazin-3-yl)piperidin-4-yl]acetic acid

  • MDL number MFCD18783092
  • Cas Number : 1279219-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186658
Availability details for
Quantity
  • Availability
  • Add to Cart